This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm04240902_s1
See other MIR1963 GE Assays ›
Gene Symbol:
Mir1963
Gene Name:
microRNA 1963
Gene Aliases:
mir-1963, mmu-mir-1963
Chromosome Location:
Chr.7: 29083636 - 29083694 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm04240902_s1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Mir1963
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_035489.1 1 30 54

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream predicted gene,
26604
Gm26604 C130030J05 -3.7kb Anti-sense Chr. 7 - 29071642 - 29079906 on NCBI Build GRCm38
Downstream RAS guanyl releasing protein 4 Rasgrp4 51.2kb Anti-sense Chr. 7 - 29134933 - 29153961 on NCBI Build GRCm38

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Mus musculus Both primers and probe map within a single exon

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0700023910.mm.1 NR_035489.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700023910.mm.1 NR_035489.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700023910.mm.1 TC0700002712.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0