This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm04238400_s1
See other MIR449B GE Assays ›
Gene Symbol:
Mir449b
Gene Name:
microRNA 449b
Gene Aliases:
Mirn449b
Chromosome Location:
Chr.13: 113037419 - 113037498 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm04238400_s1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Mir449b
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_030602.1 1 39 71

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream microRNA 449c Mir449c Mirn449b,
Mirn449c,
mir-449c
-1.3kb Sense Chr. 13 - 113035983 - 113036091 on NCBI Build GRCm38
Downstream microRNA 449a Mir449a Mirn449,
Mirn449a,
mir-449a,
mmu-mir-449
36.0 Sense Chr. 13 - 113037534 - 113037624 on NCBI Build GRCm38

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length between 71 and 85 3' Most Detect Genomic DNA Mus musculus Both primers and probe map within a single exon

Gene Ontology Categories:

Function(s)

spermatogenesis
post-embryonic development
negative regulation of gene expression
multicellular organism growth
motile cilium assembly
embryonic brain development

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1300008152.mm.1 NR_030602.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1300008152.mm.1 NR_030602.1 Refseq
Affymetrix Mouse Clariom S Array PSR1300008152.mm.1 TC1300001280.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0