This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm04238342_s1
See other MIR687 GE Assays ›
Gene Symbol:
Mir687
Gene Name:
microRNA 687
Gene Aliases:
Mirn687, mir-687, mmu-mir-687
Chromosome Location:
Chr.14: 73206763 - 73206851 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm04238342_s1
Size
Availability Made To Order
Catalog # 4426961
Price 373.00
Your Price
Online offer:
373.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Mir687
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_030459.1 1 40 73

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream retinoblastoma 1 Rb1 Rb,
Rb-1,
pRb
Sense Chr. 14 - 73194453 - 73325887 on NCBI Build GRCm38
Downstream lysophosphatidic acid receptor 6 Lpar6 2610302I02Rik,
P2ry5,
P2y5
31.0kb Anti-sense Chr. 14 - 73237891 - 73240358 on NCBI Build GRCm38

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length between 71 and 85 3' Most Detect Genomic DNA Mus musculus Both primers and probe map within a single exon

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Gene 1.1 ST Array 10421555 - -
Affymetrix Mouse Clariom S Array PSR1400018626.mm.1 NR_030459.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1400018626.mm.1 NR_030459.1 Refseq
Affymetrix Mouse Clariom S Array PSR1400018626.mm.1 TC1400002384.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0