This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm04238321_s1
See other MIR670 GE Assays ›
Gene Symbol:
Mir670
Gene Name:
microRNA 670
Gene Aliases:
Mirn670, mir-670, mmu-mir-670
Chromosome Location:
Chr.2: 94261300 - 94261399 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm04238321_s1
Size
Availability Made To Order
Catalog # 4426961
Price 373.00
Your Price
Online offer:
373.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Mir670
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_030431.1 1 53 65

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream microRNA 129-2 Mir129-2 Mirn129-2,
mir-129-2
-19.8kb Sense Chr. 2 - 94241364 - 94241453 on NCBI Build GRCm38
Downstream tetratricopeptide repeat domain 17 Ttc17 9130020K17Rik,
D2Bwg1005e
39.4kb Sense Chr. 2 - 94300766 - 94406689 on NCBI Build GRCm38

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Mus musculus Both primers and probe map within a single exon

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Gene 1.1 ST Array 10485312 - -
Affymetrix Mouse Clariom S Array PSR0200035131.mm.1 NR_030431.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200035131.mm.1 NR_030431.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200035131.mm.1 TC0200004175.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0