This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm04238320_s1
See other MIR672 GE Assays ›
Gene Symbol:
Mir672
Gene Name:
microRNA 672
Gene Aliases:
Mirn672, mir-672, mmu-mir-672
Chromosome Location:
Chr.X: 104116175 - 104116274 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm04238320_s1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Mir672
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_030430.1 1 43 83

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream expressed sequence C77370 C77370 A230051P11,
C77386,
Kiaa2022,
Xpn,
mKIAA2022
Sense Chr. X - 104077435 - 104201757 on NCBI Build GRCm38
Downstream ATP-binding cassette,
sub-family B (MDR/TAP),
member 7
Abcb7 AA517758,
AU019072,
Abc7
164.3kb Sense Chr. X - 104280565 - 104413846 on NCBI Build GRCm38

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length between 71 and 85 3' Most Detect Genomic DNA Mus musculus Both primers and probe map within a single exon

Gene Ontology Categories:

Function(s)

cellular response to lipopolysaccharide
cellular response to estrogen stimulus

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Gene 1.1 ST Array 10606215 - -
Affymetrix Mouse Clariom S Array PSR0X00017635.mm.1 NR_030430.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0X00017635.mm.1 NR_030430.1 Refseq
Affymetrix Mouse Clariom S Array PSR0X00017635.mm.1 TC0X00002795.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0