This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm04238305_s1
See other MIR301B GE Assays ›
Gene Symbol:
Mir301b
Gene Name:
microRNA 301b
Gene Aliases:
Mirn301b, mir-301b
Chromosome Location:
Chr.16: 17124400 - 17124496 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm04238305_s1
Size
Availability Made To Order
Catalog # 4426961
Price 373.00
Your Price
Online offer:
373.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Mir301b
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_030415.1 1 50 69

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream microRNA 130b Mir130b Mirn130b,
mir-130b
-258.0 Sense Chr. 16 - 17124061 - 17124142 on NCBI Build GRCm38
Downstream stromal cell-derived factor 2-like 1 Sdf2l1 5.6kb Sense Chr. 16 - 17130138 - 17132383 on NCBI Build GRCm38

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Mus musculus Both primers and probe map within a single exon

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Gene 1.1 ST Array 10438096 - -
Affymetrix Mouse Clariom S Array PSR1600010359.mm.1 NR_030415.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1600010359.mm.1 NR_030415.1 Refseq
Affymetrix Mouse Clariom S Array PSR1600010359.mm.1 TC1600001279.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0