This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm04238265_s1
See other MIR181B-2 GE Assays ›
Gene Symbol:
Mir181b-2
Gene Name:
microRNA 181b-2
Gene Aliases:
Mirn181b-2, mir-181b-2
Chromosome Location:
Chr.2: 38853830 - 38853918 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm04238265_s1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Mir181b-2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_029904.1 1 42 74
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA CJ065471.1 - 1 Chr. 2 - 38853614 - 38854226 [-] on Build GRCm38 74
GenBankmRNA CJ065621.1 - 1 Chr. 2 - 38853589 - 38854226 [-] on Build GRCm38 74

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream nuclear receptor subfamily 6,
group A,
member 1
Nr6a1 1700113M01Rik,
Gcnf,
NCNF,
RTR
Anti-sense Chr. 2 - 38723369 - 38927688 on NCBI Build GRCm38
Downstream olfactomedin-like 2A Olfml2a 4932431K08Rik,
mFLJ00237
78.1kb Sense Chr. 2 - 38931980 - 38960585 on NCBI Build GRCm38

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length between 71 and 85 3' Most Detect Genomic DNA Mus musculus Both primers and probe map within a single exon

Gene Ontology Categories:

Function(s)

negative regulation of gene expression
regulation of isotype switching
cellular response to lipopolysaccharide
cellular response to inorganic substance

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Gene 1.1 ST Array 10471880 - -
Affymetrix Mouse Clariom S Array PSR0200006258.mm.1 NR_029904.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200006258.mm.1 NR_029904.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200006258.mm.1 TC0200000712.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0