This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm04238254_s1
See other MIR217 GE Assays ›
Gene Symbol:
Mir217
Gene Name:
microRNA 217
Gene Aliases:
Mirn217, mir-217, mmu-mir-217
Chromosome Location:
Chr.11: 28763728 - 28763835 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm04238254_s1
Size
Availability Made To Order
Catalog # 4426961
Price 373.00
Your Price
Online offer:
373.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Mir217
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_029828.1 1 50 92

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream microRNA 216a Mir216a Mirn216,
Mirn216a,
mir-216a,
mmu-mir-216
-6.6kb Sense Chr. 11 - 28757012 - 28757083 on NCBI Build GRCm38
Downstream epidermal growth factor-containing fibulin-like extracellular matrix protein 1 Efemp1 89.4kb Sense Chr. 11 - 28853205 - 28926743 on NCBI Build GRCm38

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length between 86 and 100 3' Most Detect Genomic DNA Mus musculus Both primers and probe map within a single exon

Gene Ontology Categories:

Function(s)

negative regulation of gene expression
negative regulation of osteoblast differentiation
cellular response to lipopolysaccharide
cellular response to amino acid stimulus

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Gene 1.1 ST Array 10374775 - -
Affymetrix Mouse Clariom S Array PSR1100002308.mm.1 NR_029828.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100002308.mm.1 NR_029828.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100002308.mm.1 TC1100000282.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0