This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm04238234_s1
See other MIR320 GE Assays ›
Gene Symbol:
Mir320
Gene Name:
microRNA 320
Gene Aliases:
Mirn320, mir-320, mmu-mir-320
Chromosome Location:
Chr.14: 70443510 - 70443591 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm04238234_s1
Size
Availability Made To Order
Catalog # 4426961
Price 373.00
Your Price
Online offer:
373.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Mir320
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_029802.1 1 42 59

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream polymerase (RNA) III (DNA directed) polypeptide D Polr3d 2810426M17Rik,
44kDa,
AI326118,
AW489084,
BN51T,
RPC4,
TSBN51
-39.0 Anti-sense Chr. 14 - 70438748 - 70443471 on NCBI Build GRCm38
Downstream phytanoyl-CoA hydroxylase interacting protein Phyhip AW049870,
C630010D02Rik,
Lnap1ip,
PAHX-AP#1,
PAHX-AP1
13.9kb Sense Chr. 14 - 70457517 - 70468826 on NCBI Build GRCm38

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Mus musculus Both primers and probe map within a single exon

Gene Ontology Categories:

Function(s)

negative regulation of gene expression
cellular response to glucose stimulus
positive regulation of stem cell proliferation

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Gene 1.1 ST Array 10416271 - -
Affymetrix Mouse Clariom S Array PSR1400007998.mm.1 NR_029802.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1400007998.mm.1 NR_029802.1 Refseq
Affymetrix Mouse Clariom S Array PSR1400007998.mm.1 TC1400000997.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0