This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm04238216_s1
See other MIR350 GE Assays ›
Gene Symbol:
Mir350
Gene Name:
microRNA 350
Gene Aliases:
Mirn350, mir-350, mmu-mir-350
Chromosome Location:
Chr.1: 176772325 - 176772423 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm04238216_s1
Size
Availability Made To Order
Catalog # 4426961
Price 373.00
Your Price
Online offer:
373.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Mir350
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_029775.1 1 50 79

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream predicted gene,
36536
Gm36536 -245.1kb Sense Chr. 1 - 176430111 - 176527269 on NCBI Build GRCm38
Downstream serologically defined colon cancer antigen 8 Sdccag8 2700048G21Rik,
5730470G24Rik,
Cccap
42.1kb Anti-sense Chr. 1 - 176814526 - 177020437 on NCBI Build GRCm38

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length between 71 and 85 3' Most Detect Genomic DNA Mus musculus Both primers and probe map within a single exon

Gene Ontology Categories:

Function(s)

cellular response to lipopolysaccharide

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Gene 1.1 ST Array 10360504 - -
Affymetrix Mouse Clariom S Array PSR0100029761.mm.1 NR_029775.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100029761.mm.1 NR_029775.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100029761.mm.1 TC0100003622.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0