This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm04238193_s1
See other MIR31 GE Assays ›
Gene Symbol:
Mir31
Gene Name:
microRNA 31
Gene Aliases:
Mirn31, miR-31, mmu-mir-31
Chromosome Location:
Chr.4: 88910557 - 88910662 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm04238193_s1
Size
Availability Made To Order
Catalog # 4426961
Price 373.00
Your Price
Online offer:
373.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Mir31
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_029747.1 1 54 75

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream interferon epsilon Ifne Ifn-tau-1,
Ifne1,
Ifnt1,
Infe1
-30.4kb Sense Chr. 4 - 88879538 - 88880201 on NCBI Build GRCm38
Downstream predicted gene,
34568
Gm34568 45.8kb Anti-sense Chr. 4 - 88956416 - 89031301 on NCBI Build GRCm38

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length between 71 and 85 3' Most Detect Genomic DNA Mus musculus Both primers and probe map within a single exon

Gene Ontology Categories:

Function(s)

cellular response to amino acid stimulus
cellular response to inorganic substance

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Gene 1.1 ST Array 10514338 - -
Affymetrix Mouse Clariom S Array PSR0400024372.mm.1 NR_029747.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400024372.mm.1 NR_029747.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400024372.mm.1 TC0400003027.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-67c6c7f55f-2g6mz:80/100.66.128.142:80.
git-commit: 92b42c745e7842b39e6d37af9c4064bd81a92e4d
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header