This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm04238171_s1
See other MIR196A-1 GE Assays ›
Gene Symbol:
Mir196a-1
Gene Name:
microRNA 196a-1
Gene Aliases:
Mirn196, Mirn196a-1, miR-196, mir-196a-1
Chromosome Location:
Chr.11: 96265164 - 96265265 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm04238171_s1
Size
Availability Made To Order
Catalog # 4426961
Price 373.00
Your Price
Online offer:
373.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Mir196a-1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_029721.1 1 49 73

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream homeobox B9 Hoxb9 Hox-2.5 Sense Chr. 11 - 96250938 - 96276595 on NCBI Build GRCm38
Downstream homeobox B8 Hoxb8 Hox-2.4 16.6kb Sense Chr. 11 - 96281905 - 96285325 on NCBI Build GRCm38

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length between 71 and 85 3' Most Detect Genomic DNA Mus musculus Both primers and probe map within a single exon

Gene Ontology Categories:

Function(s)

mRNA cleavage
negative regulation of gene expression
Wnt signaling pathway
gene silencing by miRNA
mRNA cleavage involved in gene silencing by miRNA
cellular response to inorganic substance

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Gene 1.1 ST Array 10380620 - -
Affymetrix Mouse Clariom S Array PSR1100013808.mm.1 NR_029721.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100013808.mm.1 NR_029721.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100013808.mm.1 TC1100001491.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0