This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm04238119_s1
See other MIR137 GE Assays ›
Gene Symbol:
Mir137
Gene Name:
microRNA 137
Gene Aliases:
Mirn137, mir-137, mmu-mir-137
Chromosome Location:
Chr.3: 118433857 - 118433929 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm04238119_s1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Mir137
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_029551.1 1 38 66

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream sorting nexin 7 Snx7 2510028H01Rik -564.9kb Anti-sense Chr. 3 - 117781497 - 117868936 on NCBI Build GRCm38
Downstream predicted gene,
26871
Gm26871 70.4kb Sense Chr. 3 - 118504349 - 118528369 on NCBI Build GRCm38

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Mus musculus Both primers and probe map within a single exon

Gene Ontology Categories:

Function(s)

negative regulation of gene expression
negative regulation of osteoblast differentiation
cellular response to amino acid stimulus
cellular hyperosmotic salinity response

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Gene 1.1 ST Array 10495621 - -
Affymetrix Mouse Clariom S Array PSR0300009852.mm.1 NR_029551.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300009852.mm.1 NR_029551.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300009852.mm.1 TC0300001213.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-7cdd86b58d-kqgnb:80/100.66.128.142:80.
git-commit: 00e1f9c870be1646ae16b59a7cca7819e45b49ab
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0