This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm04238064_s1
See other RPS15A-PS6 GE Assays ›
Gene Symbol:
Rps15a-ps6
Gene Name:
ribosomal protein S15A, pseudogene 6
Gene Aliases:
Gm11968, OTTMUSG00000005107
Chromosome Location:
Chr.11: 6172508 - 6173331 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm04238064_s1
Size
Availability Made To Order
Catalog # 4426961
Price 373.00
Your Price
Online offer:
373.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Rps15a-ps6
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_029471.2 1 751 104

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream NudC domain containing 3 Nudcd3 AI427847,
BC024322,
mKIAA1068
Sense Chr. 11 - 6105691 - 6200862 on NCBI Build GRCm38
Downstream NPC1-like 1 Npc1l1 9130221N23Rik,
Gm243
37.7kb Sense Chr. 11 - 6211011 - 6230245 on NCBI Build GRCm38

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Mus musculus 1700 Both primers and probe map within a single exon

MicroArray Probes:

Agilent Affymetrix ABI

Array Name Probe IDs Targets Target Type
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2065592 NR_029471.2 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2065592 BM207059.2 mouse_transcriptome
Array Name Probe IDs Targets Target Type
Mouse Genome 430 2.0 Array 1453467_s_at - -
Mouse Genome 430 2.0 Array 1430290_at - -
Mouse Genome 430 2.0 Array 1453467_PM_s_at - -
Mouse Genome 430 2.0 Array 1430290_PM_at - -
Affymetrix GeneChip Mouse Genome 430 2.0 Array 100042173_PM_at - -
Affymetrix GeneChip Mouse Genome 430 2.0 Array 100042173_at - -
Affymetrix Mouse Clariom S Array PSR1100021067.mm.1 NR_029471.2 Refseq
Affymetrix Mouse Clariom S Array PSR1100021067.mm.1 TC1100002163.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1100021068.mm.1 NR_029471.2 Refseq
Affymetrix Mouse Clariom S Array PSR1100021068.mm.1 TC1100002163.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1100021066.mm.1 NR_029471.2 Refseq
Affymetrix Mouse Clariom S Array PSR1100021066.mm.1 TC1100002163.mm.1 MTA TC
Mouse Expression Set 430 1453467_s_at - -
Mouse Expression Set 430 1453467_PM_s_at - -
Mouse Genome 430A 2.0 Array 1453467_s_at - -
Mouse Genome 430A 2.0 Array 1453467_PM_s_at - -
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 892212 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0