This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm04238056_m1
See other GM10863 GE Assays ›
Gene Symbol:
Gm10863
Gene Name:
predicted gene 10863
Gene Aliases:
ENSMUSG00000075555
Chromosome Location:
Chr.15: 79166066 - 79216401 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm04238056_m1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Gm10863
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_029470.1 2 - 3 235 100
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA BF302801.1 - 2 - 3 235 Chr. 15 - 79166066 - 79216497 [+] on Build GRCm38 100

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream SRY (sex determining region Y)-box 10 Sox10 Dom,
Sox21
-7.5kb Anti-sense Chr. 15 - 79154909 - 79165235 on NCBI Build GRCm38
Downstream protein interacting with C kinase 1 Pick1 Prkcabp 19.1kb Sense Chr. 15 - 79229166 - 79249466 on NCBI Build GRCm38

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1500000668.mm.1

Set Membership:

Amplicon length between 86 and 100 Mus musculus Probe spans exons

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1500004994.mm.1 NR_029470.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1500004994.mm.1 NR_029470.1 Refseq
Affymetrix Mouse Clariom S Array PSR1500004994.mm.1 TC1500000668.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0