This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm04237540_m1
See other 4930562F07RIK GE Assays ›
Gene Symbol:
4930562F07Rik
Gene Name:
RIKEN cDNA 4930562F07 gene
Gene Aliases:
-
Chromosome Location:
Chr.1: 160074735 - 160077985 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm04237540_m1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: 4930562F07Rik
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_038030.1 2 - 3 237 93
NR_038031.1 2 - 3 165 93
GenBank mRNA AK016191.1 - 2 - 3 238 93
BU938572.1 - 168 93
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA BY715798.1 - 235 Chr. - - [-] on Build 93
GenBankmRNA CA466557.1 - 2 - 3 160 Chr. 1 - 160074822 - 160077977 [-] on Build GRCm38 93
GenBankmRNA BU938767.1 - 2 - 3 250 Chr. 1 - 160074970 - 160077944 [-] on Build GRCm38 93

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream tenascin R Tnr J1-tenascin,
TN-R,
janusin,
restrictin
-143.0kb Anti-sense Chr. 1 - 159523741 - 159931729 on NCBI Build GRCm38
Downstream tenascin N Tnn Tnw,
tenascin-W
7.5kb Sense Chr. 1 - 160085029 - 160153692 on NCBI Build GRCm38

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0100003349.mm.1

Set Membership:

Amplicon length between 86 and 100 Mus musculus Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0100027415.mm.1 NR_038030.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100027415.mm.1 NR_038030.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100027415.mm.1 TC0100003349.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0100027418.mm.1 NR_038031.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100027418.mm.1 NR_038031.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100027418.mm.1 TC0100003349.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0100027413.mm.1 NR_038030.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100027413.mm.1 NR_038030.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100027413.mm.1 NR_038031.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100027413.mm.1 NR_038031.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100027413.mm.1 TC0100003349.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0