This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm04236716_s1
See other G630025P09RIK GE Assays ›
Gene Symbol:
G630025P09Rik
Gene Name:
RIKEN cDNA G630025P09 gene
Gene Aliases:
4632403M07Rik
Chromosome Location:
Chr.11: 69803669 - 69806038 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm04236716_s1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: G630025P09Rik
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_027913.1 1 2155 91
GenBank mRNA AK014547.1 - 1921 91
AK036110.1 - 1794 91
AK165723.1 - 1 2156 91

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream fibroblast growth factor 11 Fgf11 Fgf-11,
Fhf-3,
Fhf3
-724.0 Anti-sense Chr. 11 - 69796068 - 69802945 on NCBI Build GRCm38
Downstream RIKEN cDNA 4933402P03 gene 4933402P03Rik 10.5kb Anti-sense Chr. 11 - 69816560 - 69818487 on NCBI Build GRCm38

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length between 86 and 100 3' Most Detect Genomic DNA Mus musculus Both primers and probe map within a single exon

MicroArray Probes:

Agilent Affymetrix

Array Name Probe IDs Targets Target Type
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2045367 NM_001033433.4 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2045367 AK143262.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2045367 BC144871.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2045367 AK165723.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2045367 AK036110.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2045367 NM_001033433.4 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2045367 AK143262.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2045367 BC144871.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2045367 AK165723.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2045367 AK036110.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P67250 AK036110.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P67250 AK014547.1 mouse_transcriptome
Array Name Probe IDs Targets Target Type
Affymetrix Mouse Gene 1.0 ST Array 10377604 - -
Affymetrix Mouse Gene 1.0 ST Array 10387649 - -
Affymetrix Mouse Gene 1.0 ST Array 10387650 - -
Affymetrix Mouse Gene 1.1 ST Array 10377603 - -
Affymetrix Mouse Gene 1.1 ST Array 10387648 - -
Affymetrix Mouse Clariom S Array PSR1100007861.mm.1 NR_027913.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100007861.mm.1 NR_027913.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100007861.mm.1 TC1100000924.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1100007863.mm.1 NR_027913.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100007863.mm.1 NR_027913.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100007863.mm.1 TC1100000924.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1100007862.mm.1 NR_027913.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100007862.mm.1 NR_027913.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100007862.mm.1 TC1100000924.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-7cdd86b58d-kqgnb:80/100.66.128.142:80.
git-commit: 00e1f9c870be1646ae16b59a7cca7819e45b49ab
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0