This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm04236415_g1
See other ZP4-PS GE Assays ›
Gene Symbol:
Zp4-ps
Gene Name:
zona pellucida glycoprotein 4, pseudogene
Gene Aliases:
EG664793, Zp4, Zpb
Chromosome Location:
Chr.13: 11522052 - 11525682 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm04236415_g1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Zp4-ps
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_027813.1 1 - 2 171 88
GenBank mRNA DQ279480.1 - 1 - 2 171 88
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA DV655471.1 - 1 - 2 69 Chr. 13 - 11522154 - 11526436 [+] on Build GRCm38 88

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream cholinergic receptor,
muscarinic 3,
cardiac
Chrm3 Chrm-3,
M3,
M3R
-1.2mb Anti-sense Chr. 13 - 9875488 - 10360803 on NCBI Build GRCm38
Downstream ryanodine receptor 2,
cardiac
Ryr2 9330127I20Rik,
RYR-2
30.5kb Anti-sense Chr. 13 - 11553099 - 12106945 on NCBI Build GRCm38

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1300000066.mm.1

Set Membership:

Amplicon length between 86 and 100 Detect Genomic DNA Mus musculus Probe spans exons

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1300000446.mm.1 NR_027813.1 Refseq
Affymetrix Mouse Clariom S Array PSR1300000446.mm.1 TC1300000066.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1300000445.mm.1 NR_027813.1 Refseq
Affymetrix Mouse Clariom S Array PSR1300000445.mm.1 TC1300000066.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0