This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm04236262_m1
See other GM20268 GE Assays ›
Gene Symbol:
Gm20268
Gene Name:
predicted gene, 20268
Gene Aliases:
-
Chromosome Location:
Chr.1: 101155768 - 101175202 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm04236262_m1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Gm20268
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_037989.1 1 - 2 245 98
GenBank mRNA AK076805.1 - 1 - 2 207 98
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA CN835055.1 - 1 - 2 188 Chr. 1 - 101155828 - 101175002 [+] on Build GRCm38 98
GenBankmRNA BU938884.1 - 1 - 2 213 Chr. 1 - 101155803 - 101175131 [+] on Build GRCm38 98
GenBankmRNA AV210161.2 - 1 - 2 236 Chr. 1 - 101155777 - 101174862 [+] on Build GRCm38 98
GenBankmRNA CA465776.1 - 196 Chr. - - [-] on Build 98
GenBankmRNA CN835259.1 - 1 - 2 248 Chr. 1 - 101155768 - 101174971 [+] on Build GRCm38 98
GenBankmRNA BB615315.1 - 1 - 2 207 Chr. 1 - 101155809 - 101174882 [+] on Build GRCm38 98

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream contactin associated protein-like 5B Cntnap5b C230078M14Rik,
Caspr5-2
-669.7kb Sense Chr. 1 - 99772247 - 100486066 on NCBI Build GRCm38
Downstream cadherin 20 Cdh20 Cdh7 3.6mb Sense Chr. 1 - 104768819 - 104995481 on NCBI Build GRCm38

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0100000989.mm.1

Set Membership:

Amplicon length between 86 and 100 Mus musculus Probe spans exons

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0100008293.mm.1 NR_037989.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100008293.mm.1 NR_037989.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100008293.mm.1 TC0100000989.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0100008292.mm.1 NR_037989.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100008292.mm.1 NR_037989.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100008292.mm.1 TC0100000989.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0