This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm04235500_m1
See other A530013C23RIK GE Assays ›
Gene Symbol:
A530013C23Rik
Gene Name:
RIKEN cDNA A530013C23 gene
Gene Aliases:
-
Chromosome Location:
Chr.2: 167690759 - 167697413 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm04235500_m1
Size
Availability Made To Order
Catalog # 4426961
Price 373.00
Your Price
Online offer:
373.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: A530013C23Rik
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_015500.3 1 - 2 85 81
GenBank mRNA AK054128.1 - 1 - 2 55 81
AK162490.1 - 1 - 2 81 81
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA BB665905.1 - 1 - 2 55 Chr. 2 - 167691208 - 167695902 [+] on Build GRCm38 81
GenBankmRNA BB636838.1 - 1 - 2 76 Chr. 2 - 167691188 - 167695797 [+] on Build GRCm38 81

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream CCAAT/enhancer binding protein (C/EBP),
beta
Cebpb C/EBPbeta,
CRP2,
IL-6DBP,
LAP,
LIP,
NF-IL6,
NF-M,
Nfil6
-745.0 Sense Chr. 2 - 167688915 - 167690432 on NCBI Build GRCm38
Downstream predicted gene 14321 Gm14321 OTTMUSG00000016308 80.2kb Sense Chr. 2 - 167777583 - 167783499 on NCBI Build GRCm38

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 3' Most Mus musculus Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0