Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix HT MG-430 PM Array Plate |
1436176_PM_at |
- |
- |
Affymetrix HT MG-430 PM Array Plate |
1429729_PM_at |
- |
- |
Affymetrix HT MG-430 PM Array Plate |
1430660_PM_at |
- |
- |
Mouse Genome 430 2.0 Array |
1430660_at |
- |
- |
Mouse Genome 430 2.0 Array |
1429729_PM_at |
- |
- |
Mouse Genome 430 2.0 Array |
1429729_at |
- |
- |
Mouse Genome 430 2.0 Array |
1436176_at |
- |
- |
Mouse Genome 430 2.0 Array |
1436176_PM_at |
- |
- |
Mouse Genome 430 2.0 Array |
1430660_PM_at |
- |
- |
Affymetrix Mouse Clariom S Array |
PSR0300017705.mm.1 |
NR_015498.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300017705.mm.1 |
TC0300002325.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0300017706.mm.1 |
NR_015498.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300017706.mm.1 |
TC0300002325.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0300017707.mm.1 |
NR_015498.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300017707.mm.1 |
TC0300002325.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0300017708.mm.1 |
NR_015498.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300017708.mm.1 |
TC0300002325.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0300017715.mm.1 |
NR_015498.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300017715.mm.1 |
TC0300002325.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0300017716.mm.1 |
NR_015498.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300017716.mm.1 |
TC0300002325.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0300017713.mm.1 |
NR_015498.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300017713.mm.1 |
TC0300002325.mm.1 |
MTA TC |
Mouse Expression Set 430 |
1436176_PM_at |
- |
- |
Mouse Expression Set 430 |
1436176_at |
- |
- |
Mouse Expression Set 430 |
1430660_at |
- |
- |
Mouse Expression Set 430 |
1430660_PM_at |
- |
- |
Mouse Expression Set 430 |
1429729_at |
- |
- |
Mouse Expression Set 430 |
1429729_PM_at |
- |
- |
Mouse Genome 430A 2.0 Array |
1436176_PM_at |
- |
- |
Mouse Genome 430A 2.0 Array |
1436176_at |
- |
- |
Affymetrix HT Mouse Genome 430A Array |
1436176_at |
- |
- |
Affymetrix HT Mouse Genome 430A Array |
1436176_PM_at |
- |
- |
Affymetrix HT Mouse Genome 430B Array |
1429729_at |
- |
- |
Affymetrix HT Mouse Genome 430B Array |
1429729_PM_at |
- |
- |
Affymetrix HT Mouse Genome 430B Array |
1430660_at |
- |
- |
Affymetrix HT Mouse Genome 430B Array |
1430660_PM_at |
- |
- |