This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm04231479_s1
See other MIR218-2 GE Assays ›
Gene Symbol:
Mir218-2
Gene Name:
microRNA 218-2
Gene Aliases:
Mirn218-2, mir-218-2
Chromosome Location:
Chr.11: 35616816 - 35616925 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm04231479_s1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Mir218-2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_029799.1 1 55 65

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream predicted gene,
42069
Gm42069 -186.6kb Sense Chr. 11 - 35426061 - 35430222 on NCBI Build GRCm38
Downstream pantothenate kinase 3 Pank3 152.6kb Sense Chr. 11 - 35769495 - 35791285 on NCBI Build GRCm38

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Mus musculus Both primers and probe map within a single exon

Gene Ontology Categories:

Function(s)

regulation of endothelial cell migration
negative regulation of gene expression
Roundabout signaling pathway
retina vasculature development in camera-type eye

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Gene 1.1 ST Array 10375214 - -
Affymetrix Mouse Clariom S Array PSR1100003137.mm.1 NR_029799.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100003137.mm.1 NR_029799.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100003137.mm.1 TC1100000381.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0