This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm04231454_s1
See other MIR139 GE Assays ›
Gene Symbol:
Mir139
Gene Name:
microRNA 139
Gene Aliases:
Mirn139, mir-139, mmu-mir-139
Chromosome Location:
Chr.7: 101475376 - 101475443 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm04231454_s1
Size
Availability Made To Order
Catalog # 4426961
Price 373.00
Your Price
Online offer:
373.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Mir139
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_029791.1 1 40 60

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream phosphodiesterase 2A,
cGMP-stimulated
Pde2a CGS-PDE,
cGSPDE
Sense Chr. 7 - 101421691 - 101512829 on NCBI Build GRCm38
Downstream predicted gene,
35363
Gm35363 45.6kb Anti-sense Chr. 7 - 101521064 - 101538664 on NCBI Build GRCm38

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Mus musculus Both primers and probe map within a single exon

Gene Ontology Categories:

Function(s)

posttranscriptional regulation of gene expression
cellular response to lipopolysaccharide

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Gene 1.1 ST Array 10555548 - -
Affymetrix Mouse Clariom S Array PSR0700012316.mm.1 NR_029791.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700012316.mm.1 NR_029791.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700012316.mm.1 TC0700001433.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0