Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1400004179.mm.1 |
NM_001146085.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400004179.mm.1 |
NM_001146085.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400004179.mm.1 |
TC1400000514.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1400004177.mm.1 |
NM_001146085.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400004177.mm.1 |
NM_001146085.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400004177.mm.1 |
NM_001146086.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400004177.mm.1 |
NM_001146086.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400004177.mm.1 |
NM_030236.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400004177.mm.1 |
NM_030236.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400004177.mm.1 |
NR_027414.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400004177.mm.1 |
NR_027414.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400004177.mm.1 |
TC1400000514.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1400004178.mm.1 |
NM_001146085.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400004178.mm.1 |
NM_001146085.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400004178.mm.1 |
NM_001146086.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400004178.mm.1 |
NM_001146086.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400004178.mm.1 |
NM_030236.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400004178.mm.1 |
NM_030236.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400004178.mm.1 |
NR_027414.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400004178.mm.1 |
NR_027414.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400004178.mm.1 |
TC1400000514.mm.1 |
MTA TC |