This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm04205641_gH
See other RPL18A GE Assays ›
Gene Symbol:
Rpl18a
Gene Name:
ribosomal protein L18A
Gene Aliases:
2510019J09Rik
Chromosome Location:
Chr.8: 70894722 - 70897443 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm04205641_gH
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Rpl18a
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_029751.4 NP_084027.1 1 - 2 88 65
GenBank mRNA AK010983.1 - 1 - 2 59 65
AK146543.1 - 1 - 2 58 65
AK150820.1 - 1 - 2 57 65
AK152611.1 - 1 - 2 57 65
AK161942.1 - 1 - 2 58 65
AK162047.1 - 1 - 2 59 65
AK168258.1 - 1 - 2 58 65
BC024609.1 - 1 - 2 45 4221597 65
BC037146.1 - 1 - 2 100 5011713 65
CJ042325.1 - 1 - 2 89 65

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the AK010983.1, AK146543.1, AK161942.1, AK162047.1, AK168258.1, BC024609.1, BC037146.1, NM_029751.4 transcripts.

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Rpl18a

Panther Classification:

Molecular Function -

ribosomal protein

Gene Ontology Categories:

Function(s) Process(es)

translation
structural constituent of ribosome
poly(A) RNA binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0800018403.mm.1 NM_029751.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR0800018403.mm.1 NM_029751.4 Refseq
Affymetrix Mouse Clariom S Array PSR0800018403.mm.1 TC0800002412.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0