This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm04205317_g1
See other MYO15 GE Assays ›
Gene Symbol:
Myo15
Gene Name:
myosin XV
Gene Aliases:
Myo15a, sh-2, sh2
Chromosome Location:
Chr.11: 60469339 - 60528369 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm04205317_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Myo15
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001103171.1 NP_001096641.1 3 - 4 3932 70
NM_010862.2 NP_034992.2 3 - 4 3932 70
NM_182698.2 NP_874357.2 2 - 3 170 70
GenBank mRNA AF053130.1 - 2 - 3 144 70
AF144095.1 - 3 - 4 3932 70
AY331132.1 - 3 - 4 3932 70
AY331133.1 - 2 - 3 170 70

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1100000755.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Myo15

Panther Classification:

Molecular Function -

actin binding motor protein

Gene Ontology Categories:

Function(s) Process(es)

sensory perception of sound
locomotory behavior
inner ear morphogenesis
nucleotide binding
motor activity
actin binding
protein binding
ATP binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1100005998.mm.1 NM_001103171.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100005998.mm.1 NM_001103171.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100005998.mm.1 NM_010862.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100005998.mm.1 NM_010862.2 Refseq
Affymetrix Mouse Clariom S Array PSR1100005998.mm.1 NM_182698.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100005998.mm.1 NM_182698.2 Refseq
Affymetrix Mouse Clariom S Array PSR1100005998.mm.1 TC1100000755.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0