This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm04203745_mH
See other IL22 GE Assays ›
Gene Symbol:
Il22 Iltifb
Gene Name:
interleukin 22
interleukin 10-related T cell-derived inducible factor beta
Gene Aliases:
IL-22, IL-22a, ILTIFa, Iltif
IL-22b, IL-TIFb, Il22b
Chromosome Location:
Chr.10: 118204586 - 118210047 on Build GRCm38
Chromosome Location:
Chr.10: 118289629 - 118295038 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
-
Assay ID Mm04203745_mH
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Il22
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_016971.2 NP_058667.1 3 - 4 425 90
GenBank mRNA BC104348.1 - 3 - 4 425 40049545 90
BC104349.1 - 3 - 4 425 40049546 90
BC116235.2 - 3 - 4 421 40091712 90
Gene Symbol: Iltifb
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_054079.2 NP_473420.2 3 - 4 447 90
GenBank mRNA AJ249492.1 - 3 - 4 90

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1000001375.mm.1, TC1000002983.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Mus musculus Inventoried Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Il22
Click here to find siRNAs for Gene Iltifb

Gene Ontology Categories:

Function(s) Process(es)

regulation of tyrosine phosphorylation of Stat3 protein
positive regulation of transcription from RNA polymerase II promoter
negative regulation of inflammatory response
reactive oxygen species metabolic process
cytokine activity
protein binding

MicroArray Probes:

Agilent Affymetrix Illumina

Array Name Probe IDs Targets Target Type
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2199202 NM_054079.2 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2199202 BC104348.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2199202 BC104349.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2199202 BC116235.2 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2199202 NM_016971.2 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2199202 NM_054079.2 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2199202 BC104348.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2199202 BC104349.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2199202 BC116235.2 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2199202 NM_016971.2 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P232628 AJ249491.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P177054 AK005228.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P177054 AJ249492.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P177054 NM_054079.2 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P177054 NM_016971.2 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P177054 AJ249491.1 mouse_transcriptome
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P09561610 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P09561613 - -
Whole Mouse Genome G4122A A_52_P177054 AK005228.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P177054 AJ249492.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P177054 NM_054079.2 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P177054 NM_016971.2 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P177054 AJ249491.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P232628 AJ249491.1 mouse_transcriptome
Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P107791 - -
Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P100960 - -
Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P09561610 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P33009512 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P32975779 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P32975765 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P32975733 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P32975770 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P32975734 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P32975788 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P33009508 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P32975778 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P32975783 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P32975767 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P32975725 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P32975758 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P33009516 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P32975736 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P32975772 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P32975784 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P32975776 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P32975781 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P32975759 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P32975786 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P32975782 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P32975761 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P32975777 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P32975774 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P32975771 - -
Array Name Probe IDs Targets Target Type
Affymetrix Mouse Gene 1.0 ST Array 10366574 - -
Affymetrix Mouse Gene 1.0 ST Array 10366578 - -
Affymetrix Mouse Gene 1.0 ST Array 10366577 - -
Affymetrix Mouse Gene 1.0 ST Array 10373699 - -
Affymetrix Mouse Gene 1.0 ST Array 10373698 - -
Affymetrix Mouse Gene 1.0 ST Array 10373696 - -
Affymetrix Mouse Gene 1.0 ST Array 10373701 - -
Affymetrix Mouse Gene 1.0 ST Array 10373697 - -
Affymetrix Mouse Gene 1.0 ST Array 10366576 - -
Affymetrix Mouse Gene 1.0 ST Array 10366575 - -
Affymetrix Mouse Gene 1.0 ST Array 10373700 - -
Affymetrix Mouse Gene 1.0 ST Array 10366573 - -
Affymetrix Mouse Gene 1.1 ST Array 10366572 - -
Affymetrix Mouse Gene 1.1 ST Array 10373695 - -
Mouse Genome 430 2.0 Array 1427624_s_at - -
Mouse Genome 430 2.0 Array 1427624_PM_s_at - -
Affymetrix Mouse Clariom S Array PSR1000022379.mm.1 NM_054079.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1000022379.mm.1 NM_054079.2 Refseq
Affymetrix Mouse Clariom S Array PSR1000022379.mm.1 TC1000002983.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1000022382.mm.1 NM_054079.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1000022382.mm.1 NM_054079.2 Refseq
Affymetrix Mouse Clariom S Array PSR1000022382.mm.1 TC1000002983.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1000022381.mm.1 NM_054079.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1000022381.mm.1 NM_054079.2 Refseq
Affymetrix Mouse Clariom S Array PSR1000022381.mm.1 TC1000002983.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1000022383.mm.1 NM_054079.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1000022383.mm.1 NM_054079.2 Refseq
Affymetrix Mouse Clariom S Array PSR1000022383.mm.1 TC1000002983.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1000010273.mm.1 NM_016971.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1000010273.mm.1 NM_016971.2 Refseq
Affymetrix Mouse Clariom S Array PSR1000010273.mm.1 TC1000001375.mm.1 MTA TC
Mouse Expression Set 430 1427624_s_at - -
Mouse Expression Set 430 1427624_PM_s_at - -
Mouse Genome 430A 2.0 Array 1427624_PM_s_at - -
Mouse Genome 430A 2.0 Array 1427624_s_at - -
Array Name Probe IDs Targets Target Type
MouseWG-6 v2 Expression BeadChip 1660102 - -
Illumina mouse-6 v1.0 expression beadchip 1660102 - -
Illumina mouseRef-8 v1.0 expression beadchip 0001660102 - -
Illumina mouseRef-8 v1.1 expression beadchip ILMN_2755950 - -
Illumina mouse-6 v1.1 expression beadchip ILMN_2755950 - -
mouse-6 v1.1 (Illumina) scl050929.1_118-S - -
Illumina Sentrix mouse-6.v1 bead array 1660102 - -
Illumina Sentrix MouseRef-8 Expression BeadChip scl050929.1_118-S - -
Illumina Sentrix Mouse-6 Expression BeadChip (2) 1660102 - -
MouseRef-8 v2 Expression BeadChip ILMN_2755950 - -
Illumina MouseWG-6 v2.0 expression beadchip ILMN_2755950 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0