This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm03978340_m1
See other D130007C19RIK GE Assays ›
Gene Symbol:
D130007C19Rik
Gene Name:
RIKEN cDNA D130007C19 gene
Gene Aliases:
-
Chromosome Location:
Chr.4: 123902072 - 123904860 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm03978340_m1
Size
Availability Made To Order
Catalog # 4426961
Price 373.00
Your Price
Online offer:
373.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: D130007C19Rik
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AK132422.1 - 57 90
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA BB570766.2 - 1 - 2 55 Chr. 4 - 123900429 - 123904841 [-] on Build GRCm38 90

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream rhomboid,
veinlet-like 2 (Drosophila)
Rhbdl2 9130416B15,
RRP2
-70.2kb Anti-sense Chr. 4 - 123787875 - 123830200 on NCBI Build GRCm38
Downstream c-myc binding protein Mycbp 5730488M09Rik,
AW552132,
AW742590,
Amy-1
172.0 Anti-sense Chr. 4 - 123905013 - 123912250 on NCBI Build GRCm38

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 3' Most Mus musculus Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0