This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm03977474_s1
See other EDARADD GE Assays ›
Gene Symbol:
Edaradd
Gene Name:
EDAR (ectodysplasin-A receptor)-associated death domain
Gene Aliases:
1810032E07Rik, 5830469M23Rik, A630043P06, cr
Chromosome Location:
Chr.13: 12471209 - 12520704 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm03977474_s1
Size
Availability Made To Order
Catalog # 4426961
Price 373.00
Your Price
Online offer:
373.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Edaradd
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_133643.4 NP_598398.3 6 5688 97
GenBank mRNA AK162653.1 - 1 3256 97

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream lectin,
galactose binding,
soluble 8
Lgals8 1200015E08Rik,
AI326142,
D13Ertd524e,
Lgals-8
-7.7kb Sense Chr. 13 - 12439402 - 12464976 on NCBI Build GRCm38
Downstream predicted gene,
30239
Gm30239 63.0kb Anti-sense Chr. 13 - 12539888 - 12562716 on NCBI Build GRCm38

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 3' Most Detect Genomic DNA Mus musculus 1700 Both primers and probe map within a single exon

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Edaradd

Panther Classification:

Molecular Function -

scaffold/adaptor protein

Gene Ontology Categories:

Function(s) Process(es)

hair follicle development
signal transduction
I-kappaB kinase/NF-kappaB signaling
multicellular organism development
cell differentiation
odontogenesis of dentin-containing tooth
trachea gland development
death receptor binding
protein binding

MicroArray Probes:

Affymetrix ABI

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1300009237.mm.1 NM_133643.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR1300009237.mm.1 NM_133643.4 Refseq
Affymetrix Mouse Clariom S Array PSR1300009237.mm.1 TC1300001449.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1300009238.mm.1 NM_133643.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR1300009238.mm.1 NM_133643.4 Refseq
Affymetrix Mouse Clariom S Array PSR1300009238.mm.1 TC1300001449.mm.1 MTA TC
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 459903 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0