This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm03977404_m1
See other LOC102640923 GE Assays ›
Gene Symbol:
LOC102640923
Gene Name:
uncharacterized LOC102640923
Gene Aliases:
-
Chromosome Location:
Chr.8: 76605226 - 76899985 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm03977404_m1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: LOC102640923
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AK158984.1 - 4 - 5 509 97
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA BY646700.1 - 1 - 2 80 Chr. 8 - 76605226 - 76797032 [-] on Build GRCm38 97

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream predicted gene 10649 Gm10649 ENSMUSG00000074235 Sense Chr. 8 - 76749087 - 76900173 on NCBI Build GRCm38
Downstream nuclear receptor subfamily 3,
group C,
member 2
Nr3c2 MR,
Mlr
102.1kb Anti-sense Chr. 8 - 76899261 - 77245232 on NCBI Build GRCm38

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 3' Most Mus musculus Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-56696975f8-skfwn:80/100.66.128.142:80.
git-commit: 795bdb1f794b4b81290d0c522fee4b91276a707f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0