This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm03976290_m1
See other GM3363 GE Assays ›
Gene Symbol:
Gm3363
Gene Name:
predicted gene 3363
Gene Aliases:
-
Chromosome Location:
Chr.2: 18742709 - 18760862 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm03976290_m1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Gm3363
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AK140973.1 - 1 - 2 287 96
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA CJ062234.1 - 1 - 2 287 Chr. 2 - 18744316 - 18760862 [-] on Build GRCm38 96

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream sperm associated antigen 6 like Spag6l -1.2kb Anti-sense Chr. 2 - 18694011 - 18749746 on NCBI Build GRCm38
Downstream phosphatidylinositol-5-phosphate 4-kinase,
type II,
alpha
Pip4k2a AW742916,
Pip5k2a
81.4kb Sense Chr. 2 - 18842255 - 18998527 on NCBI Build GRCm38

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 Mus musculus Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0