This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm03975198_m1
See other GM19673 GE Assays ›
Gene Symbol:
Gm19673
Gene Name:
predicted gene, 19673
Gene Aliases:
-
Chromosome Location:
Chr.1: 23391747 - 23399959 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm03975198_m1
Size
Availability Made To Order
Catalog # 4426961
Price 373.00
Your Price
Online offer:
373.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Gm19673
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AK090150.1 - 1 - 2 265 62
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA BE375302.1 - 1 - 2 46 Chr. 1 - 23375860 - 23397550 [-] on Build GRCm38 62
GenBankmRNA BB868666.1 - 1 - 2 266 Chr. 1 - 23392956 - 23397772 [-] on Build GRCm38 62
GenBankmRNA BB868102.1 - 1 - 2 265 Chr. 1 - 23392919 - 23397772 [-] on Build GRCm38 62
GenBankmRNA BB869912.1 - 1 - 2 267 Chr. 1 - 23392902 - 23397772 [-] on Build GRCm38 62

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream opioid growth factor receptor-like 1 Ogfrl1 2210417C17Rik -8.5kb Sense Chr. 1 - 23366424 - 23383210 on NCBI Build GRCm38
Downstream beta-1,3-glucuronyltransferase 2 (glucuronosyltransferase S) B3gat2 GlcAT-S,
Glcats
364.2kb Anti-sense Chr. 1 - 23761926 - 23847865 on NCBI Build GRCm38

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 3' Most Mus musculus Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0