This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm03972006_m1
See other LINCENC1 GE Assays ›
Gene Symbol:
Lincenc1
Gene Name:
long intergenic non-coding RNA, embryonic stem cells expressed 1
Gene Aliases:
ENSMUSG00000078952, Gm11019, Gm2373, linc-Enc1
Chromosome Location:
Chr.13: 97434970 - 97497664 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm03972006_m1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Lincenc1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_110432.1 1 - 2 116 94
GenBank mRNA BC065997.1 - 1 - 2 116 94
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA BQ044465.1 - 1 - 2 116 Chr. 13 - 97434970 - 97494952 [+] on Build GRCm38 94

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream predicted gene,
33447
Gm33447 -52.8kb Sense Chr. 13 - 97376180 - 97382156 on NCBI Build GRCm38
Downstream predicted gene 5086 Gm5086 B230311A21,
EG328314
63.0kb Sense Chr. 13 - 97560000 - 97583994 on NCBI Build GRCm38

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1300001104.mm.1

Set Membership:

Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Mus musculus Probe spans exons

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1300007058.mm.1 NR_110432.1 Refseq
Affymetrix Mouse Clariom S Array PSR1300007058.mm.1 TC1300001104.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0