Assay Details
Gene Symbol: Gm40811 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK154198.1 | - | 1 | 1151 | 98 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | fibroblast growth factor receptor substrate 2 | Frs2 |
4732458E18, C330018A15Rik, Frs2alpha, SNT1 |
-207.0 | Anti-sense | Chr. 10 - 117070127 - 117148474 on NCBI Build GRCm38 |
Downstream | RIKEN cDNA 4930423D24 gene | 4930423D24Rik | 27.6kb | Sense | Chr. 10 - 117177625 - 117181110 on NCBI Build GRCm38 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
