This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm03970739_g1
See other LOC102634154 GE Assays ›
Gene Symbol:
LOC102634154
Gene Name:
uncharacterized LOC102634154
Gene Aliases:
-
Chromosome Location:
Chr.11: 98682560 - 98685475 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm03970739_g1
Size
Availability Made To Order
Catalog # 4426961
Price 4,437.00
Your Price
Online offer:
4437.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: LOC102634154
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AK143756.1 - 37 71
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA BB843535.1 - 1 - 2 37 Chr. 11 - 98682921 - 98683415 [-] on Build GRCm38 71

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream gasdermin A Gsdma BB149167,
Gsdm,
Gsdm1,
Gsdma1,
H312E
-5.2kb Anti-sense Chr. 11 - 98664337 - 98677708 on NCBI Build GRCm38
Downstream colony stimulating factor 3 (granulocyte) Csf3 Csfg,
G-CSF,
MGI-IG
17.7kb Anti-sense Chr. 11 - 98701095 - 98704836 on NCBI Build GRCm38

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.

Set Membership:

Amplicon length between 71 and 85 3' Most Detect Genomic DNA Mus musculus Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0