This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm03965187_m1
See other LOC101055751 GE Assays ›
Gene Symbol:
LOC101055751
Gene Name:
uncharacterized LOC101055751
Gene Aliases:
-
Chromosome Location:
-
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm03965187_m1
Size
Availability Made To Order
Catalog # 4426961
Price 373.00
Your Price
Online offer:
373.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: LOC101055751
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AK042120.1 - 1 - 2 44 59
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA BB638927.1 - 1 - 2 44 Chr. 2 - 34912871 - 34914789 [+] on Build GRCm38 59

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream cutA divalent cation tolerance homolog-like Cutal D730039F16Rik -20.7kb Sense Chr. 2 - 34874365 - 34892133 on NCBI Build GRCm38
Downstream TNF receptor-associated factor 1 Traf1 4732496E14Rik 27.2kb Anti-sense Chr. 2 - 34943256 - 34961772 on NCBI Build GRCm38

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 3' Most Mus musculus Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-7c5f86c6b-2xgzq:80/100.66.128.142:80.
git-commit: c124ca8d7fcef91bd439783ab4009544f5b84403
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0