This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm03961803_m1
See other 4930486F22RIK GE Assays ›
Gene Symbol:
4930486F22Rik
Gene Name:
RIKEN cDNA 4930486F22 gene
Gene Aliases:
-
Chromosome Location:
Chr.10: 86095854 - 86113586 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm03961803_m1
Size
Availability Made To Order
Catalog # 4426961
Price 373.00
Your Price
Online offer:
373.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: 4930486F22Rik
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AK133288.1 - 50 60
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA BB616668.1 - 1 - 2 50 Chr. 10 - 86095839 - 86127120 [+] on Build GRCm38 60

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream synapsin III Syn3 Anti-sense Chr. 10 - 86048746 - 86499019 on NCBI Build GRCm38
Downstream tissue inhibitor of metalloproteinase 3 Timp3 Timp-3 174.5kb Sense Chr. 10 - 86300412 - 86349505 on NCBI Build GRCm38

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Mus musculus Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0