This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm03961731_m1
See other LOC102632231 GE Assays ›
Gene Symbol:
LOC102632231
Gene Name:
uncharacterized LOC102632231
Gene Aliases:
-
Chromosome Location:
Chr.15: 6581170 - 6652522 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm03961731_m1
Size
Availability Made To Order
Catalog # 4426961
Price 373.00
Your Price
Online offer:
373.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: LOC102632231
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AK133228.1 - 308 98
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA BB616548.1 - 2 - 3 308 Chr. 15 - 6619745 - 6654305 [-] on Build GRCm38 98

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream FYN binding protein Fyb ADAP,
B630013F22Rik,
FYB-120/130,
Slap130,
p120/130,
p120/p130
Anti-sense Chr. 15 - 6522797 - 6665608 on NCBI Build GRCm38
Downstream RPTOR independent companion of MTOR,
complex 2
Rictor 4921505C17Rik,
6030405M08Rik,
AVO3,
AW492497,
D530039E11Rik
55.8kb Anti-sense Chr. 15 - 6708349 - 6800401 on NCBI Build GRCm38

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 Mus musculus Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0