This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm03959157_m1
See other 9030205G03RIK GE Assays ›
Gene Symbol:
9030205G03Rik
Gene Name:
RIKEN cDNA 9030205G03 gene
Gene Aliases:
-
Chromosome Location:
-
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm03959157_m1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: 9030205G03Rik
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AK020245.1 - 1 - 2 452 76
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA BY720642.1 - 1 - 2 452 Chr. 8 - 14381732 - 14507248 [+] on Build GRCm38 76

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream discs,
large (Drosophila) homolog-associated protein 2
Dlgap2 6430596N04Rik,
DAP2,
Sapap2
Sense Chr. 8 - 14094615 - 14848626 on NCBI Build GRCm38
Downstream ceroid-lipofuscinosis,
neuronal 8
Cln8 mnd 373.6kb Sense Chr. 8 - 14881335 - 14901720 on NCBI Build GRCm38

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 3' Most Mus musculus Probe spans exons

MicroArray Probes:

Agilent Affymetrix Illumina

Array Name Probe IDs Targets Target Type
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2199737 AK020245.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2199737 AK020245.1 mouse_transcriptome
Array Name Probe IDs Targets Target Type
Affymetrix HT MG-430 PM Array Plate 1432241_PM_at - -
Mouse Genome 430 2.0 Array 1432241_at - -
Mouse Genome 430 2.0 Array 1432241_PM_at - -
Mouse Expression Set 430 1432241_at - -
Mouse Expression Set 430 1432241_PM_at - -
Affymetrix HT Mouse Genome 430B Array 1432241_PM_at - -
Affymetrix HT Mouse Genome 430B Array 1432241_at - -
Array Name Probe IDs Targets Target Type
MouseWG-6 v2 Expression BeadChip 101780017 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-7c5f86c6b-2xgzq:80/100.66.128.142:80.
git-commit: c124ca8d7fcef91bd439783ab4009544f5b84403
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0