This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm03958704_s1
See other 9430099H24RIK GE Assays ›
Gene Symbol:
9430099H24Rik
Gene Name:
RIKEN cDNA 9430099H24 gene
Gene Aliases:
-
Chromosome Location:
-
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm03958704_s1
Size
Availability Made To Order
Catalog # 4426961
Price 373.00
Your Price
Online offer:
373.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: 9430099H24Rik
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AK020529.1 - 1 854 69
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA BB106222.1 - 1 127 Chr. 3 - 86268377 - 86268680 [+] on Build GRCm38 69

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream ribosomal protein S3A1 Rps3a1 Rps3a -125.0kb Anti-sense Chr. 3 - 86137940 - 86142668 on NCBI Build GRCm38
Downstream mab-21-like 2 (C. elegans) Mab21l2 276.9kb Anti-sense Chr. 3 - 86545581 - 86548283 on NCBI Build GRCm38

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Mus musculus Both primers and probe map within a single exon

MicroArray Probes:

Agilent Affymetrix

Array Name Probe IDs Targets Target Type
Whole Mouse Genome G4122A A_51_P450074 AK020529.1 mouse_transcriptome
Array Name Probe IDs Targets Target Type
Affymetrix HT MG-430 PM Array Plate 1454363_PM_at - -
Affymetrix HT MG-430 PM Array Plate 1432806_PM_at - -
Mouse Genome 430 2.0 Array 1432806_at - -
Mouse Genome 430 2.0 Array 1454363_at - -
Mouse Genome 430 2.0 Array 1432806_PM_at - -
Mouse Genome 430 2.0 Array 1454363_PM_at - -
Mouse Expression Set 430 1432806_PM_at - -
Mouse Expression Set 430 1454363_PM_at - -
Mouse Expression Set 430 1432806_at - -
Mouse Expression Set 430 1454363_at - -
Affymetrix HT Mouse Genome 430B Array 1432806_PM_at - -
Affymetrix HT Mouse Genome 430B Array 1454363_PM_at - -
Affymetrix HT Mouse Genome 430B Array 1432806_at - -
Affymetrix HT Mouse Genome 430B Array 1454363_at - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0