This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm03958571_s1
See other 9430076D03RIK GE Assays ›
Gene Symbol:
9430076D03Rik
Gene Name:
RIKEN cDNA 9430076D03 gene
Gene Aliases:
-
Chromosome Location:
-
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm03958571_s1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: 9430076D03Rik
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AK020488.1 - 1 50 83
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA BY721851.1 - 1 50 Chr. 16 - 38527732 - 38528416 [+] on Build GRCm38 83

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream protein O-glucosyltransferase 1 Poglut1 9630046K23Rik,
Clp46,
Ktelc1,
Rumi
Anti-sense Chr. 16 - 38525058 - 38550256 on NCBI Build GRCm38
Downstream transmembrane protein 39a Tmem39a 2610033C09Rik 30.0kb Sense Chr. 16 - 38558698 - 38592162 on NCBI Build GRCm38

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length between 71 and 85 3' Most Detect Genomic DNA Mus musculus 1700 Both primers and probe map within a single exon

MicroArray Probes:

ABI Illumina

Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 851756 - -
Array Name Probe IDs Targets Target Type
MouseWG-6 v2 Expression BeadChip 102650035 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-7cdd86b58d-kqgnb:80/100.66.128.142:80.
git-commit: 00e1f9c870be1646ae16b59a7cca7819e45b49ab
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0