This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm03958445_s1
See other 9430024C03RIK GE Assays ›
Gene Symbol:
9430024C03Rik
Gene Name:
RIKEN cDNA 9430024C03 gene
Gene Aliases:
-
Chromosome Location:
-
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm03958445_s1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: 9430024C03Rik
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AK020434.1 - 1 850 111

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream UTP20,
small subunit (SSU) processome component,
homolog (yeast)
Utp20 3830408P06Rik,
AA617408,
Drim,
mDRIM,
mDRIN
-47.8kb Sense Chr. 10 - 88746607 - 88826814 on NCBI Build GRCm38
Downstream solute carrier family 5 (iodide transporter),
member 8
Slc5a8 Ait,
SMCT
10.2kb Anti-sense Chr. 10 - 88885992 - 88929515 on NCBI Build GRCm38

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Mus musculus Both primers and probe map within a single exon

MicroArray Probes:

Agilent Affymetrix

Array Name Probe IDs Targets Target Type
Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P101957 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27327852 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27327856 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P08314883 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27327858 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P08314874 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P08314877 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27327859 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27327853 - -
Array Name Probe IDs Targets Target Type
Affymetrix HT MG-430 PM Array Plate 1433216_PM_at - -
Mouse Genome 430 2.0 Array 1433216_PM_at - -
Mouse Genome 430 2.0 Array 1433215_at - -
Mouse Genome 430 2.0 Array 1433216_at - -
Mouse Genome 430 2.0 Array 1433215_PM_at - -
Mouse Expression Set 430 1433215_PM_at - -
Mouse Expression Set 430 1433215_at - -
Mouse Expression Set 430 1433216_at - -
Mouse Expression Set 430 1433216_PM_at - -
Affymetrix HT Mouse Genome 430B Array 1433216_PM_at - -
Affymetrix HT Mouse Genome 430B Array 1433216_at - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-56696975f8-skfwn:80/100.66.128.142:80.
git-commit: 795bdb1f794b4b81290d0c522fee4b91276a707f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0