This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm03958406_s1
See other 9430019C24RIK GE Assays ›
Gene Symbol:
9430019C24Rik
Gene Name:
RIKEN cDNA 9430019C24 gene
Gene Aliases:
-
Chromosome Location:
-
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm03958406_s1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: 9430019C24Rik
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AK020424.1 - 1 954 105
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA BM121183.2 - 1 Chr. 8 - 48378507 - 48379146 [+] on Build GRCm38 105
GenBankmRNA BQ557441.1 - 1 Chr. 8 - 48378507 - 48379006 [+] on Build GRCm38 105

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream teneurin transmembrane protein 3 Tenm3 2610100B16Rik,
Odz1,
Odz3,
Ten-m3,
mKIAA1455
Sense Chr. 8 - 48225665 - 48935162 on NCBI Build GRCm38
Downstream predicted gene,
19744
Gm19744 464.2kb Anti-sense Chr. 8 - 48843885 - 48870497 on NCBI Build GRCm38

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length greater than or equal to 101 Detect Genomic DNA Mus musculus Both primers and probe map within a single exon

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0