This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm03958371_s1
See other 9430006E15RIK GE Assays ›
Gene Symbol:
9430006E15Rik
Gene Name:
RIKEN cDNA 9430006E15 gene
Gene Aliases:
-
Chromosome Location:
-
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm03958371_s1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: 9430006E15Rik
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AK020405.1 - 1 846 95

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream olfactomedin 2 Olfm2 A030009A06Rik Anti-sense Chr. 9 - 20667691 - 20746311 on NCBI Build GRCm38
Downstream collagen,
type V,
alpha 3
Col5a3 81.2kb Anti-sense Chr. 9 - 20770050 - 20815117 on NCBI Build GRCm38

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length between 86 and 100 3' Most Detect Genomic DNA Mus musculus 1700 Both primers and probe map within a single exon

MicroArray Probes:

Affymetrix ABI

Array Name Probe IDs Targets Target Type
Affymetrix HT MG-430 PM Array Plate 1454589_PM_at - -
Affymetrix HT MG-430 PM Array Plate 1454588_PM_at - -
Mouse Genome 430 2.0 Array 1454589_at - -
Mouse Genome 430 2.0 Array 1454589_PM_at - -
Mouse Genome 430 2.0 Array 1454588_PM_at - -
Mouse Genome 430 2.0 Array 1454588_at - -
Mouse Expression Set 430 1454588_PM_at - -
Mouse Expression Set 430 1454588_at - -
Mouse Expression Set 430 1454589_at - -
Mouse Expression Set 430 1454589_PM_at - -
Affymetrix HT Mouse Genome 430B Array 1454589_at - -
Affymetrix HT Mouse Genome 430B Array 1454588_at - -
Affymetrix HT Mouse Genome 430B Array 1454588_PM_at - -
Affymetrix HT Mouse Genome 430B Array 1454589_PM_at - -
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 639983 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0