This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm03958257_s1
See other 9230110I02RIK GE Assays ›
Gene Symbol:
9230110I02Rik
Gene Name:
RIKEN cDNA 9230110I02 gene
Gene Aliases:
-
Chromosome Location:
-
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm03958257_s1
Size
Availability Made To Order
Catalog # 4426961
Price 373.00
Your Price
Online offer:
373.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: 9230110I02Rik
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AK020330.1 - 1 854 75

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream B cell CLL/lymphoma 9 Bcl9 2610202E01Rik,
8030475K17Rik,
A330041G23Rik,
Gm130
Sense Chr. 3 - 97203658 - 97298083 on NCBI Build GRCm38
Downstream olfactory receptor 1402 Olfr1402 MOR217-1 138.6kb Sense Chr. 3 - 97410217 - 97411179 on NCBI Build GRCm38

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length between 71 and 85 3' Most Detect Genomic DNA Mus musculus Both primers and probe map within a single exon

MicroArray Probes:

Agilent Affymetrix

Array Name Probe IDs Targets Target Type
Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P139601 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P139601 - -
Whole Mouse Genome G4122A A_51_P356536 AK020330.1 mouse_transcriptome
Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P140290 - -
Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P139601 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P22353740 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P22353729 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P02538770 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P22353722 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P22353693 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P22353694 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P22353692 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P22353732 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P02516726 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P22353698 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P22353721 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P03124867 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P03031222 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P02739334 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P22353743 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P03031229 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P22353733 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P02394637 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P22353759 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P22353758 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P22353762 - -
Array Name Probe IDs Targets Target Type
Affymetrix HT MG-430 PM Array Plate 1460499_PM_at - -
Mouse Genome 430 2.0 Array 1460499_at - -
Mouse Genome 430 2.0 Array 1460499_PM_at - -
Mouse Expression Set 430 1460499_PM_at - -
Mouse Expression Set 430 1460499_at - -
Affymetrix HT Mouse Genome 430B Array 1460499_at - -
Affymetrix HT Mouse Genome 430B Array 1460499_PM_at - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0