This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm03957739_s1
See other ZFP950 GE Assays ›
Gene Symbol:
Zfp950
Gene Name:
zinc finger prtoein 950
Gene Aliases:
5830428H23Rik, AU016073, BC055757, Gm34518, Zfp826, mszf84
Chromosome Location:
Chr.19: 61051147 - 61313507 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm03957739_s1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Zfp950
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AK020020.1 - 1 1745 99
AK164548.1 - 3 4190 100
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA BG088303.2 - 1 135 Chr. 19 - 61122657 - 61123236 [-] on Build GRCm38 100
GenBankmRNA BG229383.1 - 1 142 Chr. 19 - 61122949 - 61123242 [-] on Build GRCm38 100

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream G protein-coupled receptor kinase 5 Grk5 Gprk5 -26.5kb Anti-sense Chr. 19 - 60889593 - 61095968 on NCBI Build GRCm38
Downstream predicted gene 6020 Gm6020 EG547091,
mszf72
27.0kb Anti-sense Chr. 19 - 61151864 - 61184658 on NCBI Build GRCm38

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length between 86 and 100 3' Most Detect Genomic DNA Mus musculus Both primers and probe map within a single exon

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0