This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm03957617_s1
See other 5730585A16RIK GE Assays ›
Gene Symbol:
5730585A16Rik
Gene Name:
RIKEN cDNA 5730585A16 gene
Gene Aliases:
-
Chromosome Location:
-
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm03957617_s1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: 5730585A16Rik
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AK019974.1 - 1 854 95

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream predicted gene,
30528
Gm30528 Sense Chr. 15 - 103026805 - 103063780 on NCBI Build GRCm38
Downstream Riken cDNA D930007P13 gene D930007P13Rik 63.8kb Sense Chr. 15 - 103123070 - 103146828 on NCBI Build GRCm38

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length between 86 and 100 3' Most Detect Genomic DNA Mus musculus Both primers and probe map within a single exon

MicroArray Probes:

Agilent

Array Name Probe IDs Targets Target Type
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1969087 AK019974.1 mouse_transcriptome
Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P104712 - -
Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P147214 - -
Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P128102 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P128102 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P104712 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P147214 - -
Whole Mouse Genome G4122A A_51_P450007 AK019974.1 mouse_transcriptome
Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P101546 - -
Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P147214 - -
Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P128102 - -
Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P104712 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P11917223 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30501807 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30501841 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P12311699 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30501846 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30501852 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30501835 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30501815 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30501870 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30501808 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30501810 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30501829 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30501879 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P11917221 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30501838 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30501827 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30501861 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30501837 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P11917217 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P11838461 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P12180014 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P11863360 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30501876 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P12284180 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P11838450 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30501824 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30501814 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30501881 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30501832 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P12180019 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30501819 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P12284173 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P12206082 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30501817 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30501844 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30501885 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30501850 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30501873 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30501816 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P11838446 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30501848 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P12160797 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30501857 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30501863 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30501853 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30501839 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30501856 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P12125519 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-7cdd86b58d-kqgnb:80/100.66.128.142:80.
git-commit: 00e1f9c870be1646ae16b59a7cca7819e45b49ab
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0