Assay Details
Gene Symbol: 4930567J20Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_040576.1 | 2 - 3 | 486 | 80 | ||
GenBank mRNA | AK019789.1 | - | 2 - 3 | 486 | 80 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | RIKEN cDNA 4931420L22 gene | 4931420L22Rik | -120.5kb | Sense | Chr. 16 - 71131076 - 71139029 on NCBI Build GRCm38 | |
Downstream | roundabout homolog 1 (Drosophila) | Robo1 |
AW494633, AW742721, DUTT1, Gm310 |
760.4kb | Sense | Chr. 16 - 72027553 - 73047491 on NCBI Build GRCm38 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
Agilent
Affymetrix
ABI
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 822 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 470562 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 584006 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 17585 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P03223139 | - | - |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P107729 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P03223139 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30854084 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30854133 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30854110 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30854116 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30854147 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30854156 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30854140 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30854096 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30854158 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30854107 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30854160 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30854188 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30854130 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30854142 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30854127 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30854150 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30854221 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30854163 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30854196 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30854179 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30854114 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30854207 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30854159 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30854217 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30854112 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30854092 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30854152 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30854190 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30854146 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30854115 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30854220 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30854197 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30854088 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30854215 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30854198 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30854204 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P12629175 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30854193 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30854086 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30854166 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30854125 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30854195 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30854213 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P12650541 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30854175 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30854091 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30854184 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30854090 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30854138 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30854165 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30854206 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P12847276 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30854218 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30854108 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30854211 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30854168 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30854209 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30854173 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P12963027 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30854223 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P12711082 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P12731549 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30854095 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30854151 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30854143 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30854119 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P12806422 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30854102 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30854132 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30854136 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30854123 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30854144 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30854194 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30854172 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30854186 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30854224 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30854121 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30854082 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30854135 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30854104 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30854089 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30854100 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30854201 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30854170 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30854177 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P12845417 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30854222 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30854178 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30854185 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30854226 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P12617316 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30854203 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Clariom S Array | PSR1600006918.mm.1 | NR_040576.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1600006918.mm.1 | NR_040576.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1600006918.mm.1 | TC1600000818.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1600006919.mm.1 | NR_040576.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1600006919.mm.1 | NR_040576.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1600006919.mm.1 | TC1600000818.mm.1 | MTA TC |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 768260 | - | - |
