This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm03957397_s1
See other 4930563H03RIK GE Assays ›
Gene Symbol:
4930563H03Rik
Gene Name:
RIKEN cDNA 4930563H03 gene
Gene Aliases:
-
Chromosome Location:
-
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm03957397_s1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: 4930563H03Rik
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AA178695.1 - 1 326 96
AK019780.1 - 1 246 96
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA BY715814.1 - 1 246 Chr. 15 - 79074706 - 79075048 [-] on Build GRCm38 96
GenBankmRNA BX637579.1 - 1 Chr. 15 - 79074714 - 79075050 [+] on Build GRCm38 96
GenBankmRNA AI451052.1 - 1 Chr. 15 - 79074719 - 79075091 [+] on Build GRCm38 96

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream ankyrin repeat domain 54 Ankrd54 C730048E16Rik,
EST1068184,
EST475269,
Liar
-11.8kb Sense Chr. 15 - 79053094 - 79062958 on NCBI Build GRCm38
Downstream eukaryotic translation initiation factor 3,
subunit L
Eif3l 0610011H21Rik,
D15N1e,
Eif3eip,
Eif3ip,
Eif3s6ip,
HSP-66Y,
PAF67
175.0 Anti-sense Chr. 15 - 79075223 - 79094400 on NCBI Build GRCm38

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length between 86 and 100 3' Most Detect Genomic DNA Mus musculus Both primers and probe map within a single exon

MicroArray Probes:

Agilent Affymetrix Illumina

Array Name Probe IDs Targets Target Type
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 917964 - -
Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P118478 - -
Array Name Probe IDs Targets Target Type
Murine Genome U74v2 Set 141023_at - -
Murine Genome U74v2 Set 141023_PM_at - -
Array Name Probe IDs Targets Target Type
MouseWG-6 v2 Expression BeadChip 103990372 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-6f7678f454-w8g9k:80/100.66.128.142:80.
git-commit: 7b0558793d0e70b0085ba95ba375cdfe261f8cbf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0