This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm03957287_s1
See other D130043K22RIK GE Assays ›
Gene Symbol:
D130043K22Rik
Gene Name:
RIKEN cDNA D130043K22 gene
Gene Aliases:
4930451E12Rik, Kiaa0319
Chromosome Location:
Chr.13: 24843930 - 24902159 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm03957287_s1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: D130043K22Rik
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AK019624.1 - 1 454 68
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA BY714986.1 - 1 450 Chr. 13 - 24845115 - 24845724 [+] on Build GRCm38 68

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream tyrosyl-DNA phosphodiesterase 2 Tdp2 D13Ertd656e,
Ttrap
-3.0kb Sense Chr. 13 - 24831647 - 24842153 on NCBI Build GRCm38
Downstream RIKEN cDNA 4932702P03 gene 4932702P03Rik 1700047I16Rik 59.9kb Sense Chr. 13 - 24905600 - 24911780 on NCBI Build GRCm38

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Mus musculus Both primers and probe map within a single exon

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-7cdd86b58d-kqgnb:80/100.66.128.142:80.
git-commit: 00e1f9c870be1646ae16b59a7cca7819e45b49ab
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0